• USA Home
  • Z374695 - Eppendorf® Combitips® Plus Biopur®
EMAIL THIS PAGE TO A FRIEND
Z374695 Sigma
Eppendorf® Combitips® Plus Biopur®
volume range 1.0 mL
Purchase
Properties
Related Categories Combitips Plus Biopur, Eppendorf, Eppendorf PCR Pipette Tips, Eppendorf Pipette Tips, Eppendorf Pipettors & Tips,
material   colorless tip
sterility   sterile
feature   ATP free
  DNA free (Bacterial (E. Coli))
  DNA free (Human)
  DNase free
  PCR inhibitor free
  RNase free
  barrier: no
  pyrogen free
packaging   pack of 100
mfr. no.   Eppendorf®, 0030 069.439 (European)
  Eppendorf®, 0030 069.439
  Eppendorf®, 022496069 (USA)
volume range   1.0 mL
Description
General description
Snaps securely into Multipette pipette. Tips are not autoclavable. Biopur tips are certified sterile and pyrogen, RNase-, DNA and ATP-free.
Legal Information
Biopur is a registered trademark of Eppendorf AG
Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?