• USA Home
  • Z374822 - Stovall hybridization bottle carousel
EMAIL THIS PAGE TO A FRIEND
Z374822 Sigma
Stovall hybridization bottle carousel
Universal carousel
Purchase
Description
General description
Optional carousel holds four - 55 × 170 or 210 mm acrylic bottles
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?