• USA Home
  • Z374954 - Pierceable cap strips For PCR tubes
EMAIL THIS PAGE TO A FRIEND
Z374954 Sigma
Pierceable cap strips For PCR tubes
Purchase
Properties
packaging   pkg of 120 strips (960 caps)
Description
General description
Caps in strips of eight; the center of each cap can be pierced with a hypodermic needle for quick sample removal without generating aerosols or other sources of cross-contamination. Caps can be used with 0.2 mL PCR strip tubes and 96-well plates.
Price and Availability
Your Key to Success
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Protocols
Long and Accurate PCR Amplification of DNA with RedAccuTaq D4812 Protocol
Reaction Optimization Reliable amplification of long DNA sequences requires: 1) effective denaturation of DNA template, 2) adequate extension times to produce large products and 3) protection of targ...
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?