• USA Home
  • Z375039 - Vertical electrophoresis unit, fixed height, single gel
EMAIL THIS PAGE TO A FRIEND
Z375039 Sigma
Vertical electrophoresis unit, fixed height, single gel
gel W × H 16.5 cm × 14.5 cm, CE-approved
Purchase
Description
Features and Benefits
• Made from high quality acrylic with platinum electrodes
• Long lasting silicone sponge gasket on upper reservoir for leak-free operation without grease
• Safety covers with attached safety-tip power leads
• Designed for use with unique gel-casting gasket; heavy-duty casting clamps compress gasket between glass plates, sealing without grease
For Slab Gel Electrophoresis:
• Designed to use as little as 200 mL of buffer, and as little as 5 μg of sample per well
• Use with gel casting accessories which offer the ability to cast one or two gels in a single assembly
• Use in standard slab gel electrophoresis or in the second dimension of 2D electrophoresis
For Tube Gel Electrophoresis:
• Only the two fixed-height dual-gel units accept tube gel accessories
• Tubes in four different inside diameters all have the same outside diameter so they are interchangeable in the tube gel adapter
• Two-sided operation allows up to 20 tubes to be run at a time
Each unit comes with base unit, upper and lower reservoirs with safety covers and attached power leads, and two gel-mounting spring clamps.
CE APPROVED ONLY WHERE INDICATED
Slab and tube gel accessories are not included
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?