• USA Home
  • Z375144 - Masterflex pump system for aseptic filling
EMAIL THIS PAGE TO A FRIEND
Z375144 Sigma
Masterflex pump system for aseptic filling
with precision standard drive, AC input 90 - 130 V
Purchase
Properties
mfr. no.   Cole Parmer
availability   not available in EU
Description
Features and Benefits
• Separate single-turn speed control and on/off switch with green power indicator. Maintain speed setting while turning on/off.
• The 1/20-hp unidirectional motor has soft start and back EMF for ±5% speed control. Smooth operation and long service life.
• Stackable painted steel housing; IP22 rated. Protects drive from falling water.
• 230 VAC models: complies with CE regulations.
• 6-ft (1.8-m) line cord—230 VAC: IEC 320/CEE22 socket.
• Drive accepts eight different pump head types: Standard, Easy-Load, Easy-Load II, Quick Load, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm.
General description
A perfect upgrade from a fixed-speed drive, these variable-speed drives include a wider rpm range, ±5% speed regulation, and UL, cUL, and CE listings. For added versatility, these drives accept eight different pump head types.
Drives and heads can be purchased separately. All drives are UL, cUL, and CE compliant; model is supplied with a Euro plug. A UK power cord is available separately (Z375349).
Includes C-Flex ULTRA L/S 24 tubing 06434-24 (10 ft)
Legal Information
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?