• USA Home
  • Z375276 - Masterflex pump drive
EMAIL THIS PAGE TO A FRIEND
Z375276 Sigma
Masterflex pump drive
economy drive, AC input 90 - 130 V
Purchase
Properties
Related Categories Labware, Peristaltic Pumps, Pumps More...
feature   CSA approved
mfr. no.   Cole Parmer
availability   not available in EU
Description
General description
Does not include pump head.
Economy:
unidirectional, lowest priced variable-speed drive, ± 5% speed uniformity. Accepts one EASY-LOAD head
Precision: bidirectional, soft start, 10-turn speed control, ± 1% speed uniformity. Accepts DB9 cable for remote control capabilities
Digital: bidirectional, high-precision digital speed control, ± 0.25% speed uniformity. Displays rpm, flow rate, and % speed. Accepts DB9 cable for remote control capabilities
Legal Information
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?