• USA Home
  • Z375292 - Masterflex Easy-Load peristaltic pump head
EMAIL THIS PAGE TO A FRIEND
Z375292 Sigma
Masterflex Easy-Load peristaltic pump head
Easy-Load L/S for high-performance precision tubing
Purchase
Properties
Related Categories Labware, Peristaltic Pumps, Pumps More...
material   stainless steel rotor
mfr. no.   Cole Parmer
Description
General description
One of these heads is included with the Masterflex pump systems for aseptic filling. Accepts two high-performance precision tubing sizes that carry 1.7-2.8 ml/rev. This tubing has thicker walls enabling better pressure generation, suction lift, pumping of viscous fluids, and longer tubing service.
Legal Information
Easy-Load is a trademark of Cole-Parmer Instrument Co.
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?