• USA Home
  • Z375322 - Masterflex Easy-Load peristaltic pump head
EMAIL THIS PAGE TO A FRIEND
Z375322 Sigma
Masterflex Easy-Load peristaltic pump head
PTFE diaphragm, pump head
Purchase
Properties
Related Categories Labware, Peristaltic Pumps, Pumps More...
material   PTFE diaphragm
mfr. no.   Cole Parmer
Description
General description
All wetted parts are PTFE, making this the ideal head for demanding applications:
• Unaffected by strong acids, alkali, most organic solvents
• Handles liquids up to 65 °C (continuous); 121 °C (intermittent)
• Pressures to 50 psi (continuous); 75 psi (intermittent)
• Low rates from 10-800 mL/min with most 600 rpm Masterflex drives (not compatible with Economy drive).
Inlet and outlet ports are standard 1/4 in. outside thread NPT, and are equipped with check valves to maintain prime and prevent siphoning.
Legal Information
Easy-Load is a trademark of Cole-Parmer Instrument Co.
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?