• USA Home
  • Z375772 - Circulators refrigerated/heated
EMAIL THIS PAGE TO A FRIEND
Z375772 Sigma
Circulators refrigerated/heated
Model 9105, digital, AC input 240 V AC
Purchase
Properties
mfr. no.   Polyscience
availability   not available in EU
Description
Features and Benefits
These circulators feature a 6-liter access reservoir in a space saving vertical design. Each unit has a LED readout, redundant safety backup, and a sealed compressor for years of maintenance-free use. Additionally, the units are equipped with a 1/4′′ NPT internally threaded pump inlet and outlet for external circulation. Model911 is analog design, with a temperature range of −20 to 100 °C ±2 °C, and requires thermometer temperature readout. Model9105 has a digital display, readable in °C or °F with an accuracy of ±0.5 °C, and can be set from −20 to 150 °C. All models use CFC refrigerant and have over-temp and low-liquid cut-offs.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?