EMAIL THIS PAGE TO A FRIEND
Z375896 Sigma
Shaker platforms
AC/DC input 120 V
Purchase
Properties
mfr. no.   Polyscience
availability   not available in EU
Description
Features and Benefits
The multi-purpose shaker platform transforms the 20-liter bath into an incubator/shaker. Dual action design allows for both reciprocal and orbital shaking action. Stroke is adjustable between 2-12 mm, and speed from 30 to 180 RPM.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?