EMAIL THIS PAGE TO A FRIEND
Z375926 Sigma
Shaker lid
pkg of 1 ea
Purchase
Properties
Related Categories Baths and Circulators, Julabo Baths for Flammable Liquids, Labware, Manufacturer Browser, Polyscience More...
packaging   pkg of 1 ea
mfr. no.   Polyscience
availability   not available in EU
Description
Features and Benefits
Specially designed to cover samples while allowing for the shaker¢s tray arms. The lid is steeply gabled to allow for flasks up to 1000 mL.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?