• USA Home
  • Z375934 - Plastic floating balls for water bath
EMAIL THIS PAGE TO A FRIEND
Z375934 Sigma
Plastic floating balls for water bath
pkg of 400
Purchase
Properties
Related Categories Baths and Circulators, General Purpose Baths and Circulators, Labware, Manufacturer Browser, Polyscience More...
material   polypropylene
packaging   pkg of 400
mfr. no.   Polyscience
Description
General description
These hollow polypropylene balls reduce evaporation and help maintain temperature within the water bath.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?