• USA Home
  • Z375950 - Electroporator, EC100
EMAIL THIS PAGE TO A FRIEND
Z375950 Sigma
Electroporator, EC100
AC input 240 V AC
Purchase
Properties
Related Categories Electroporation, Labware, Manufacturer Browser, Savant
availability   not available in EU
Description
Features and Benefits
The EC100 electroporator is compact, versatile, and economical. Three optimized preset voltages make this unit perfect for prokaryotic and eukaryotic transformations. Just load and insert the disposable cuvette, close the safety cover, select the setting, and push the red button. An audible and visible signal indicates that the pulse has been delivered. With a recharge time of less than 10 seconds, the unit will be ready again by the time you′ve loaded the next cuvette.
Recommended voltage/cuvette combinations:
• Mammalian: 400 V, 4-mm cuvette = 1.0 kV/cm
• Bacteria: 1800 V, 1-mm cuvette = 18 kV/cm or 2800 V, 2-mm cuvette = 14 kV/cm
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?