• USA Home
  • Z375985 - Electroporation system, Cell-Porator®
EMAIL THIS PAGE TO A FRIEND
Z375985 Sigma
Electroporation system, Cell-Porator®
Voltage booster, AC input 120 V
Purchase
Properties
availability   not available in EU
Description
Compatibility
Use with optional voltage booster for high-field pulses, up to 16.6kV/cm.
Features and Benefits
This system provides a method of introducing foreign proteins or nucleic acids into prokaryotic and eukaryotic cells with convenience, safety, and complete control. Chamber safe holds four electroporation chambers for sequential processing. The Cell-Porator itself maintains the sample temperature during processing. The pulse control unit delivers a 400V pulse, which can be set for durations from <0.1 to >200 msec. An internal shunt resistor can be switched in or out for high- or low-salt cell suspensions. System also includes a chamber rack and 36 disposable electroporation chambers with 4 mm gap (provides 1kV/cm for mammalian and plant cell transformations).
Legal Information
Cell-Porator is a registered trademark of Gibco BRL Life Technologies, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?