• USA Home
  • Z376019 - Electroporation chambers
EMAIL THIS PAGE TO A FRIEND
Z376019 Sigma
Electroporation chambers
0.15 cm gap
Purchase
Properties
availability   not available in EU
Description
Features and Benefits
Specifically designed for the Cell-Porator system and available with electrode gap widths of 1.5 mm (for bacterial and yeast) or 4 mm (for mammalian and plant cells)
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?