• USA Home
  • Z376051 - Spectronic standards kit
EMAIL THIS PAGE TO A FRIEND
Z376051 Sigma
Spectronic standards kit
Purchase
Properties
Related Categories Labware, Spectroscopy Supplies, Spectroscopy, UV More...
Description
General description
For certified evaluation of the performance and accuracy of the instrument in your own laboratory. Set contains standards for 0% transmittance, stray radiant energy (at 3 wavelengths), certified wavelength (at 3 wavelengths) and photometric performance (accuracy, linearity, precision and optical alignment). Kit is complete with cleaning brush, storage case and manual.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?