• USA Home
  • Z376248 - Gel Repel glass plate coating
EMAIL THIS PAGE TO A FRIEND
Z376248 Sigma
Gel Repel glass plate coating
Purchase
Description
General description
A non-toxic alternative to silanization for pre-treatment of glass plates for sequencing or other PAGE techniques. Pre-treatment assures trouble-free disassembly of gel sandwich.
Packaging
Each cloth will treat one or two sequencing plates or several PAGE plates (depending on size).
Legal Information
Gel Repel is a trademark of C.B.S. Scientific Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z719951 Gel Repel liquid solution, contains 4 × 1 oz. vials and instructions
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?