EMAIL THIS PAGE TO A FRIEND
Z376256 Sigma
Gel dryer, EC355
AC input 120 V, 60 Hz
Purchase
Properties
availability   not available in EU
Description
General description
With a 40 × 50 cm drying platform, the EC355 Gel Dryer is big enough for large sequencing gels or several smaller gels at once. Digital programming of temperature and time ensure reproducible drying, and LEDs indicate status throughout the run. At the end of the program, the AUTO mode turns off heat and the gel cools under vacuum. There is also a HOLD mode that keeps heat and vacuum applied until manually terminated. The drying surface is inert PTFE, and vapors flow through a corrosion resistant path, assuring dependable operation.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?