• USA Home
  • Z376337 - Shaker flask holders for Polyscience shaker sample tray
EMAIL THIS PAGE TO A FRIEND
Z376337 Sigma
Shaker flask holders for Polyscience shaker sample tray
250 mL flasks
Purchase
Properties
Related Categories Baths and Circulators, General Purpose Baths and Circulators, Labware, Manufacturer Browser, Polyscience More...
material   stainless steel
sterility   non-sterile
packaging   pack of 1
mfr. no.   Polyscience
availability   not available in EU
capacity   250 mL
Description
Application
Easily mount to the shaker sample tray to secure flasks.
Compatibility
Fit flasks from 25 mL to 1000 ml
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?