• USA Home
  • Z376361 - Gel dryer (EC355) and pump (EC353) accessory
EMAIL THIS PAGE TO A FRIEND
Z376361 Sigma
Gel dryer (EC355) and pump (EC353) accessory
Chemical trapping kit
Purchase
Description
Features and Benefits
Specifically designed for trapping radioactive residues pulled from gel during drying. Includes trap, tubing, connectors, and disposable cartridge (dispose properly as radioactive waste).
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?