• USA Home
  • Z376434 - miniPERM® classic kits
EMAIL THIS PAGE TO A FRIEND
Z376434 Sigma
miniPERM® classic kits
MWCO 12.5 kDa
Purchase
Properties
Related Categories Cell Culture Supplies, Greiner, Greiner Culture Supplies, Greiner miniPERM, Labware,
mfr. no.   Greiner
pore size   12.5 kDa MWCO
Description
General description
An economical, easy-to-use bioreactor which simplifies the high density production of hybridomas and other cell types,. Also enables the cost efficient production of high purity monoclonal antibodies and other extracellular materials (interleukins, viruses, recombinant proteins and peptides). The system consists of two modules separated by a dialysis membrane. The membrane prevents the passage of cells and high molecular weight molecules.
Nutrients, products of cell metabolism and gases diffuse through the membrane. Consequently the products secreted by the cells will be producted in higher concentration than possible in conventional stationary tissue culture.
Depending on the characteristics of the cell line, densities in excess of 107 cells/ml and antibody concentrations of several mg per mL are possible.
Can be used with other bottle turning devices, but for maximum production, bottle turners capable of 40 rpm are required.
Packaging
Each kit contains:
• 4 reusable nutrient modules
• 8 spare screw caps for nutrient modules
• 12 single-use, sterile production modules
• 36 spare screw caps for production modules
• 4 stands for the miniPERM units
• 1 opener
Legal Information
miniPERM is a registered trademark of Heraeus Instruments GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?