• USA Home
  • Z376493 - miniPERM® HDC 5 Kit
EMAIL THIS PAGE TO A FRIEND
Z376493 Sigma
miniPERM® HDC 5 Kit
MWCO 12.5 kDa
Purchase
Properties
Related Categories Cell Culture Supplies, Greiner, Greiner Culture Supplies, Greiner miniPERM, Labware,
mfr. no.   Greiner
pore size   12.5 kDa MWCO
Description
Features and Benefits
A 5 mL module especially designed to cultivate to high densities in a shorter time. Available with MWCO of 3.5 kDa or 12.5 kDa.
Packaging
Each kit contains:
• 4 reusable nutrient modules
• 8 screw caps for nutrient module
• 12 sterile, single-use production modules
• 36 screw caps for production module
• 4 miniPERM stands
• 1 opener
Legal Information
miniPERM is a registered trademark of Heraeus Instruments GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?