• USA Home
  • Z376612 - miniPERM® silicone fingers with support frames and one opener
EMAIL THIS PAGE TO A FRIEND
Z376612 Sigma
miniPERM® silicone fingers with support frames and one opener
Purchase
Properties
packaging   package of 2
mfr. no.   Greiner
Description
Application
Silicone fingers are used to facilitate efficient oxygen transfer to support high density culturing.
Legal Information
miniPERM is a registered trademark of Heraeus Instruments GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?