• USA Home
  • Z376620 - miniPERM® bottle turning device
EMAIL THIS PAGE TO A FRIEND
Z376620 Sigma
miniPERM® bottle turning device
Bottle roller, AC/DC input 230 V AC
Purchase
Properties
mfr. no.   Greiner
Description
General description
Designed to fit in most incubators. Suitable for 4 miniPERM bioreactors. Speed is selectable between 0-40 rpm in increments of 0.1 rpm.
Legal Information
miniPERM is a registered trademark of Heraeus Instruments GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?