EMAIL THIS PAGE TO A FRIEND
Z376655 Sigma
flexiPERM® discs
hydrophobic, silicone
Purchase
Properties
Related Categories Cell Culture Supplies, Culture Dishes, Greiner, Greiner Culture Dishes, Greiner Culture Supplies,
material   silicone
description   Fits 50 mm Lumox culture dish (Z376744)
packaging   pkg of 5
mfr. no.   Greiner
chamber diam. × H × volume   15 mm × 9 mm × 500-800 μL
Description
General description
Reusable inserts divide the units into 4 quadrants. Heat and cold resistant and not toxic to cells.
Legal Information
flexiPERM is a registered trademark of Heraeus Instruments GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?