• USA Home
  • Z376663 - quadriPERM® accessories
EMAIL THIS PAGE TO A FRIEND
Z376663 Sigma
quadriPERM® accessories
flexiPERM® Micro 12
Synonym: chamber slide, chamber slides
Purchase
Description
General description
A reusable silicone product that divides a quadriPERM chamber into 12 cell culture vessels. The micro12 is heat- and cold-resistant, hydrophobic and non-toxic to cells.
Legal Information
flexiPERM is a registered trademark of Heraeus Instruments GmbH
quadriPERM is a registered trademark of Heraeus Instruments GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?