• USA Home
  • Z376744 - Greiner Lumox culture dish
EMAIL THIS PAGE TO A FRIEND
Z376744 Sigma
Greiner Lumox culture dish
size 50 mm, hydrophilic
Synonym: culture dishes, tissue culture dishes, tissue culture plates
Purchase
Properties
Related Categories Cell Culture Supplies, Culture Dishes, Greiner, Greiner Culture Dishes, Greiner Culture Supplies,
sterility   sterile
packaging   pkg of 5
mfr. no.   Greiner 9607 7303
size   50 mm
Description
General description
Previously known as petriPERM culture dishes, this dish has a gas-permeable base allowing cells to grow faster at the gas/liquid interface. With ordinary plates, the medium forms a barrier to gas diffusion, limiting cell growth. The base-membrane is crystal clear, UV transparent and does not fluoresce. Cells grown on the membrane can be stained in place or the membrane can be cut for staining, documentation or other applications.
Legal Information
Lumox is a trademark of Greiner Bio-One GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?