• USA Home
  • Z376760 - quadriPERM® cell culture vessel
EMAIL THIS PAGE TO A FRIEND
Z376760 Sigma
quadriPERM® cell culture vessel
rectangular in shape and split into four separate chambers
Synonym: cell culture system, cell culture systems
Purchase
Description
General description
A rectangular cell culture vessel that is split into four separate chambers for standard sized microscope slides. The quadriPERM allows for parallel growth of cells under identical conditions either on the surface of the vessel or on coated or uncoated slides. Cells can be fixed, stained, or tagged and are suitable for all molecular genetic and immunological detection processes. The units are gamma irradiated and designed for single use.
Legal Information
quadriPERM is a registered trademark of Heraeus Instruments GmbH
Price and Availability
Pharma QC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?