• USA Home
  • Z376841 - Dispet, Nichiryo
EMAIL THIS PAGE TO A FRIEND
Z376841 Sigma
Dispet, Nichiryo
size 1.0-5.0 mL
Purchase
Properties
availability   not available in EU
size   1.0-5.0 mL
Description
General description
A variable volume bottle top dispenser with accuracy of better than ±1% and precision of ±0.2%. All models will fit reagent bottles with 27 mm threads. The 0.2-1 mL dispenser is supplied with a 22 mm adaptor. The dispensing mechanism is completely solvent- and corrosion-proof, permitting use with all liquids except hydrofluoric acid. Ceramic plunger (1 mL and 5 mL), and Pyrex glass plunger (10 mL) assure smooth action.
Legal Information
Dispet is a trademark of Nichiryo Co. Ltd.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?