• USA Home
  • Z376876 - Nichiryo digital hand-held dispenser
EMAIL THIS PAGE TO A FRIEND
Z376876 Sigma
Nichiryo digital hand-held dispenser
0.2-1.0 mL dispensing volume
Purchase
Properties
availability   not available in EU
measuring range   0.2-1.0 mL dispensing volume
Description
General description
This light weight dispenser allows for continuous dispensing of solutions. Glass cylinder and plunger are protected from breakage by plastic housings. All parts which come in direct contact with the solution are autoclavable. Provides accuracy of ±1% and precision of <0.2%.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?