EMAIL THIS PAGE TO A FRIEND
Z376930 Sigma
Pipette sheath
Fits up to 250 μl pipetters
Purchase
Properties
sterility   sterile
packaging   pkg of 100
mfr. no.   Diversified Biotech
Description
General description
Reduce the risk of cross-contamination in critical pipetting applications like PCR. Nuclease-free disposable sheaths slip over the pipette barrel after the tip has been placed. Forms a seamless cover that protects samples as well as pipette from contamination. Sheath ejects along with pipette tip (on pipetters with tip ejectors).
Packaging
Individually wrapped
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?