• USA Home
  • Z376949 - Glowtape luminescent darkroom labelling tape
EMAIL THIS PAGE TO A FRIEND
Z376949 Sigma
Glowtape luminescent darkroom labelling tape
Purchase
Properties
roll W × L   1 in. × 500 in.
Description
Application
Use to mark drawers, equipment, tools, solutions in darkrooms. After brief exposure to room light, tape will glow for up to 20 minutes. Write on tape with ordinary black marker for easily readable darkroom labels. Glow will not affect film unless brought in close proximity or left for prolonged periods. Tape is washable and resistant to most darkroom chemicals.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?