• USA Home
  • Z376965 - Acculab® GS Balance
EMAIL THIS PAGE TO A FRIEND
Z376965 Sigma
Acculab® GS Balance
Model GS-200
Purchase
Properties
material   plastic weighing plate
mfr. no.   Acculab
availability   not available in EU
measuring range   200 g weight, resolution: 0.1 g, linearity: 0.2 g
  7 oz weight, resolution: 0.01 oz
platform diam.   13.3x13.3 cm
Description
General description
Economy toploader GS balances feature:
• weighing in grams or avoirdupois ounces
• tare to capacity
• auto-OFF in battery-powered mode
• LCD with overload, underload, and low battery indicators
• factory preset calibration
• plastic housing; plastic platform 13.3 × 13.3cm
Legal Information
Acculab is a registered trademark of Globaltec Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?