• USA Home
  • Z377198 - Peristaltic pump tubing Nalgene® 50
EMAIL THIS PAGE TO A FRIEND
Z377198 Sigma
Peristaltic pump tubing Nalgene® 50
For L/S 24 pump head
Purchase
Properties
Related Categories Labware, Peristaltic Pumps, Pumps More...
mfr. no.   Nalgene
Description
General description
Flexible, translucent and durable, Nalgene 50 high purity silicone tubing is designed for a variety of pump and transfer applications including pharmaceutical, laboratory, and bioprocess manufacturing . There are no phthalate plasticizers that can leach out into transported fluid. It also resists adhesion of tissue, blood, proteins, and food products. Manufactured under strict process controls for precise dimensional tolerances. Can be autoclaved for 30 minutes at 121 °C (15 psi) in muslin cloth or sterilizing paper. Non-cytotoxic, non-pyrogenic and non-hemolytic. NOTE: not intended for invasive use.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?