• USA Home
  • Z377260 - Peristaltic pump tubing Nalgene® 570
EMAIL THIS PAGE TO A FRIEND
Z377260 Sigma
Peristaltic pump tubing Nalgene® 570
For L/S 15 pump head
Purchase
Properties
mfr. no.   Nalgene
Description
General description
Nalgene 570 Biologically Pure Tubing (BPT) is equivalent to Norton PharMed. It meets Food Grade USP Class VI and non-cytotoxic requirements. Offers superior chemical resistance. Sterilizable by autoclave, dry heat, or gas.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?