• USA Home
  • Z377309 - Peristaltic pump tubing Nalgene® 570
EMAIL THIS PAGE TO A FRIEND
Z377309 Sigma
Peristaltic pump tubing Nalgene® 570
For L/S 17 pump head
Purchase
Properties
Related Categories Labware, Peristaltic Pumps, Pumps More...
mfr. no.   Nalgene
Description
General description
Nalgene 570 Biologically Pure Tubing (BPT) is equivalent to Norton PharMed. It meets Food Grade USP Class VI and non-cytotoxic requirements. Offers superior chemical resistance. Sterilizable by autoclave, dry heat, or gas.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?