• USA Home
  • Z377392 - Bottles Nalgene® style 1630
EMAIL THIS PAGE TO A FRIEND
Z377392 Sigma
Bottles Nalgene® style 1630
size 125 mL
Synonym: nalgene bottle, nalgene bottles
Purchase
Properties
material   PTFE
description   Autoclavable
mfr. no.   Nalgene
size   125 mL
Description
General description
These narrow-mouth bottles are among the best temperature resistant and chemical resistant containers available. Leakproof.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?