• USA Home
  • Z377430 - Nalgene® FastCap bottle-top filter unit
EMAIL THIS PAGE TO A FRIEND
Z377430 Sigma
Nalgene® FastCap bottle-top filter unit
pore size 0.45 μm
Purchase
Properties
sterility   sterile
feature   cap plug seal
mfr. no.   Nalgene
filter size   90 mm
pore size   0.45 μm
Description
General description
Filter from the prep flask directly into the receiver. These bottle-top filters with low protein binding PES membrane have a universal FastCap fitting. Just place it on the mouth of the receiver and apply vacuum. Filters up to 5 liters. Sterilized by γ-irradiation.
Legal Information
FastCap is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?