• USA Home
  • Z377562 - The Belly Button® mini-orbital shaker
EMAIL THIS PAGE TO A FRIEND
Z377562 Sigma
The Belly Button® mini-orbital shaker
AC input 230 V AC50 - 60 Hz, European 2-pin plug
Purchase
Properties
Related Categories IBI Scientific, IBI Scientific Shakers, Labware, Manufacturer Browser, Shakers & Vortexers,
feature   CE compliant
  0 ‑ 8 ° platform pitch control
mfr. no.   IBI
parameter   0-100 rpm speed
W × D × H   8 in. × 8 in. × 5 in.
platform W × L   7 in. × 7 in.
Description
Frequently Asked Questions
Frequently Asked Questions are available for this Product.
General description
The Belly Button is a compact version of the well known Belly Dancer shaker. Designed with available space in mind, the Belly Button has a compact 8 in.x 8in. foot print and an over all height of only 5 inches. The easily adjustable speed and platform pitch angle controles provide a wide range of agitation conditions, from extremely gentle to vigorous.
The Belly Button is an exceptionally simple liquid agitation system, consisting of orbiting platform flexibly anchored at four corners so that the platform motion, when adjusted by the platform pitch control collar, is an extremely effective combination of rocking platform and orbital shaker. One-dimensional shakers do not have the versatility and range of application that result from the variable speed and adjustable platform pitch of the Belly Button.
Ideal to use in a crowded incubator for call culture and tissue culture applications.
To use this product in the UK please order Z768588 separately
Legal Information
The Belly Button is a registered trademark of IBI Scientific Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Frequently Asked Questions
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
What are the specifications/dimensions for Product Z377562, Belly Button™ mini-orbital shaker?
The Belly Button™ mini-orbital shaker, Product Z377562, specifications/dimensions are: Dimensions    8" x 8" x 4 ¾" h Platform    7" x7" Weight    5 lbs. Ambient Temp Operation    0°C to 60°C Max Load    5 lbs. Electrical    Universal voltage, 50 / 60 hz Motor    Brushless DC Speed    0 to 100 rpm @ 115v / 230v ac
Show more questions
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?