• USA Home
  • Z377627 - Polaroid®, Macro 5 SLR camera
EMAIL THIS PAGE TO A FRIEND
Z377627 Sigma
Polaroid®, Macro 5 SLR camera
Purchase
Description
Features and Benefits
An instant, close-up system that is easy to use, versatile, and portable. Offers 5 magnifications (.2×, .4×, 1×, 2×, 3×) at the turn of a dial. The micro-processor-controlled exposure system automatically corrects exposure and the dual electronic flash provides even illumination at all magnifications. Uses Spectra Type 990 Polaroid film (F 7772).
Legal Information
Polaroid is a registered trademark of Polaroid Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?