• USA Home
  • Z377953 - ABI DNA Sequencing Unit Accessories
EMAIL THIS PAGE TO A FRIEND
Z377953 Sigma
ABI DNA Sequencing Unit Accessories
Casting Comb, thickness 0.2 mm × 1 well
Purchase
Description
General description
Sigma offers glass plates, Mylar combs, and spacers designed specifically to meet the rigid specifications of the ABI 373 and 377 DNA sequencing units. Notched glass places are beveled for easy handling; well-to-read (WTR) distances as well as overall dimensions are given. Combs have printed well numbers and center marks. Glass plates can be purchased individually or in sets.
Special features of Step Plates:
• Front plate is beveled to create a thicker gel in the comb area, giving enough sample capacity for 96 lanes
• Gel casting is standard, but uses a 0.4 mm casting comb and sharktooth comb
Special features of Microtrough 96 plates:
• Microtroughs notched inside the front plate provide enough sample capacity for 96 lanes
• A special Mylar plug instead of a standard casting comb
• Microtroughs provide greater loading area and guide your pipette tops for more accurate sample placement, eliminating the need for expensive loaders or specialized tips
Legal Information
ABI is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?