• USA Home
  • Z378704 - Labpette® digital multichannel pipette
EMAIL THIS PAGE TO A FRIEND
Z378704 Sigma
Labpette® digital multichannel pipette
8 channel, size 20-200 μL
Purchase
Properties
Related Categories Labnet, Labnet Pipettors, Labnet Pipettors & Tips, Labware, Liquid Handling Equipment,
feature   electronic: no
mfr. no.   Labnet
availability   not available in EU
size   20-200 μL
Description
General description
A new generation of precision microliter pipettes. Light-weight, well balanced, comfortable to use even for prolonged repetitive pipetting. Self-adjusting, self-lubricating seals and finely polished stainless steel piston provide superior accuracy and reproducibility. Eight- and twelve-channel pipettes feature continuously adjustable, digital micrometers for volume selection. Each channel has its own finely polished stainless steel piston to ensure reproducibility from one pipetting series to the next as well as from channel to channel. The instrument is light in weight and well balanced, with a slender ribbed hand grip for comfortable use with right or left hand. Tip ejector can be conveniently activated by thumb or index finger.
Legal Information
Labpette is a registered trademark of National Labnet
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?