• USA Home
  • Z379166 - CoverWell perfusion chambers
EMAIL THIS PAGE TO A FRIEND
Z379166 Sigma
CoverWell perfusion chambers
4 chambers per slide, chamber oblong, volume 70 μL
Purchase
Properties
Related Categories Labware, Light Microscopy, Microscopy & Staining More...
material   chamber oblong
mfr. no.   Grace Bio-Labs
D × L × W   0.5 mm × 19 mm × 6 mm
volume   70 μL
Description
General description
CoverWell perfusion chambers are press-to-seal covers which form multiple cell incubation or cytochemistry chambers when pressed to glass coverslips or microscope slides. Reagents can be quickly added or removed through dual access ports without disturbing or cross-contaminating specimens. Suitable for live cell imaging on upright or inverted, visible light and fluorescence microscopes.
Legal Information
CoverWell is a trademark of Grace Bio-Labs, Inc.
Price and Availability
Tissue Diagnostics
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
GBL622504 Grace Bio-Labs CoverWell perfusion chambers, 4 chambers, adhesive size 0.5 mm, ports diam. 1.5 mm, chamber W × L × thickness 6.35 mm × 19 mm × 0.6 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?