• USA Home
  • Z379263 - SequeSoap glass plate cleaner

EMAIL THIS PAGE TO A FRIEND
Z379263 Sigma

SequeSoap glass plate cleaner

Purchase

Description

Application

Removes contaminants from glass plates used in nucleic acid sequencing. Strips away residues of glass coating agents that can build up and interfere with optimal separations. Compatible with hand and machine washing. A 2% solution is recommended as a starting concentration (somewhat less for machine washing to control foaming). Phosphate-free and biodegradable.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?