• USA Home
  • Z379476 - Hanna general purpose combination pH electrode
EMAIL THIS PAGE TO A FRIEND
Z379476 Sigma
Hanna general purpose combination pH electrode
Ultem body (Double junction refillable)
Purchase
Properties
Related Categories Hanna, Hanna general purpose combination pH electrode, Hanna pH Electrodes, Hanna pH Meters and Accessories, Labware,
description   BNC connector
  Double junction Ag/AgCI
mfr. no.   Hanna HI 1332B
measuring range   0-14 pH
  0-80 °C temp
L   120 mm , Overall
Description
General description
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
60137 Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl
replaced by
Product #
Description
Add to Cart
Z276456 Orion® Ross® Sure-Flow® electrode, Test tube
Z276448 Orion® Ross® Sure-Flow® electrode, Rugged
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?