• USA Home
  • Z380628 - Hanna Piccolo® pH meter
EMAIL THIS PAGE TO A FRIEND
Z380628 Sigma
Hanna Piccolo® pH meter
replacement electrode for Piccolo 2
Purchase
Properties
Related Categories Hand Held, Hanna, Hanna Piccolo pH meter, Hanna pH Meters, Hanna pH Meters and Accessories,
feature   calibration manual dual point, ±1 pH
  temperature compensation 0 ‑ 70 °C (automatic with integral temperature sensor)
mfr. no.   HI1290
measuring range   0-14 pH, accuracy: 0.01, resolution: 0.01
L   160 mm
weight   400 gincluding calibration screwdriver, ABS carrying case, and batteries
  70 g
battery power   3 × 1.4 V button cell, 100 hr life (continuous operation)
Description
General description
Hand-held stick-style pH/temperature meters with rugged design for field use. "Amplified electrode" delivers strong signal to the meter, eliminating the vulnerability to noise, humidity, and dirt that is characteristic of conventional meters.
Legal Information
Piccolo is a registered trademark of Hanna Instruments
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?