• USA Home
  • Z380652 - SequeStrip glass plate cleaner

EMAIL THIS PAGE TO A FRIEND
Z380652 Sigma

SequeStrip glass plate cleaner

Purchase

Description

Application

Regenerates the surface of glass electrophoresis plates by removing dirt and minor imperfections and scratches. Plates should then be washed with SequeSoap.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
FK6310000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?