Z381284 Sigma
Bridge mounting plate for Techne® immersion circulator
for 8-26 liter baths
DOWNLOAD MSDS (PDF)
Description
Compatibility
Required for use of TE-10A, TE-10D, and TU-20D thermoregulators; included with TU-20C
Legal Information
Techne is a registered trademark of Techne, Inc.
Price and Availability
Customers Also Viewed
Techne<SUP>®</SUP> immersion circulator
Techne TE-10A, AC input 120 V
Techne<SUP>®</SUP> immersion circulator
Techne TE-10D, AC input 120 V
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?