• USA Home
  • Z381373 - Techne® Dri-Block® heater
EMAIL THIS PAGE TO A FRIEND
Z381373 Sigma
Techne® Dri-Block® heater
Model DB-3, AC/DC input 240 V AC
Purchase
Properties
Related Categories Block Heaters, Heating, Labware, Manufacturer Browser, Techne,
usage   three insert blocks
feature   CE compliant
  calibrated analogue dial (±2°C).
mfr. no.   Techne
measuring range   25-100 °C temperature
Description
Components
Supplied with both EU (2-pin) & GB (3-pin) power cords.
General description
Three different models offer a selection of temperature ranges and features, and accept a wide variety of insert blocks for your specific needs. All are sturdily constructed of aluminum and engineering-grade plastic with all solid-state controls, and are provided with a block removal tool. Efficient insulation keeps case cool even at maximum operating temperature. Pilot lights indicate power on/off, heater action, and over-temperature warning.
Heating blocks/inserts not supplied (order separately)
Legal Information
Dri-Block is a registered trademark of Techne, Inc.
Techne is a registered trademark of Techne, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?