• USA Home
  • Z381381 - Techne® Dri-Block® heater
EMAIL THIS PAGE TO A FRIEND
Z381381 Sigma
Techne® Dri-Block® heater
Model DB-3D, AC input 120 V
Purchase
Properties
Related Categories Block Heaters, Heating, Labware, Manufacturer Browser, Techne,
usage   three insert blocks
feature   CSA approved: No
  dial and set button (±1°C) with digital LED display.
mfr. no.   Techne
availability   not available in EU
measuring range   25-200 °C temperature
Description
General description
Accepts up to three insert blocks; temperature range: 25-200°C; settings via dial and set button (±1°C) with digital LED display.
Three different models offer a selection of temperature ranges and features, and accept a wide variety of insert blocks for your specific needs. All are sturdily constructed of aluminum and engineering-grade plastic with all solid-state controls, and are provided with a block removal tool. Efficient insulation keeps case cool even at maximum operating temperature. Pilot lights indicate power on/off, heater action, and over-temperature warning.
Heating blocks/inserts not supplied (order separately)
Legal Information
Dri-Block is a registered trademark of Techne, Inc.
Techne is a registered trademark of Techne, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?